accessibility menu, dialog, popup

UserName

CAS RN 1135-40-6

IUPAC Name: 3-(cyclohexylamino)propane-1-sulfonic acid
Synonyms: caps (buffer) CAPS
Molecular Weight (g/mol): 221.32
Molecular Formula: C9H19NO3S
InChi Key: PJWWRFATQTVXHA-UHFFFAOYSA-N
SMILES: OS(=O)(=O)CCCNC1CCCCC1
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (4)
Physical Form 
  • (10)
  • (3)
  • (2)
pH 
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (12)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (4)
  • (10)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (10)
  • (2)
  • (3)
  • (3)
  • (3)