accessibility menu, dialog, popup

UserName

CAS RN 1134-76-5

H3CNCH3OH2N

CAS RN 1134-76-5

IUPAC Name: 2-[3-(dimethylamino)propoxy]aniline
Synonyms: 2-[3-(dimethylamino)propoxy]aniline
Molecular Weight (g/mol): 194.28
Molecular Formula: C11H18N2O
InChi Key: XGTIDBFMHOJXSS-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results