accessibility menu, dialog, popup

UserName

CAS RN 1134-50-5

OONN

CAS RN 1134-50-5

IUPAC Name: 1-phenyl-1H-pyrazole-4-carboxylate
Synonyms: 1-phenylpyrazole-4-carboxylate
Molecular Weight (g/mol): 187.18
Molecular Formula: C10H7N2O2
InChi Key: ZROILLPDIUNLSE-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CN(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results