accessibility menu, dialog, popup

UserName

CAS RN 1134-49-2

OHONHN

CAS RN 1134-49-2

IUPAC Name: 3-phenyl-1H-pyrazole-5-carboxylic acid
Synonyms: 5-phenyl-2H-pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: QBPUOAJBMXXBNU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NN1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results