accessibility menu, dialog, popup

UserName

CH3FOClF

CAS RN 112857-70-2

IUPAC Name: 2,4-difluoro-3-methylbenzoyl chloride
Synonyms: 2,4-difluoro-3-methylbenzoyl chloride
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: BMMDDDOEASRDKX-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(C(Cl)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results