accessibility menu, dialog, popup

UserName

CAS RN 112811-66-2

H3COFFOClF

CAS RN 112811-66-2

IUPAC Name: 2,4,5-trifluoro-3-methoxybenzoyl chloride
Synonyms: 2,4,5-trifluoro-3-methoxybenzoyl chloride
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: JVQSZTJTWSUJCR-UHFFFAOYSA-N
SMILES: COC1=C(F)C(=CC(F)=C1F)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results