CAS RN 112758-40-4
CAS RN 112758-40-4
IUPAC Name:
5-methyl-1H-pyrazole-4-carbaldehyde
Synonyms:
3-methyl-2H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol):
110.12
Molecular Formula:
C5H6N2O
InChi Key:
NWDMGTFNIOCVDU-UHFFFAOYSA-N
SMILES:
CC1=C(C=O)C=NN1
1
–
1
of
1
results
1
3-Methyl-1H-pyrazole-4-carboxaldehyde, 97%
CAS: 112758-40-4 Molecular Formula: C5H6N2O Molecular Weight (g/mol): 110.12 MDL Number: MFCD01313822 InChI Key: NWDMGTFNIOCVDU-UHFFFAOYSA-N Synonym: 3-methyl-1h-pyrazole-4-carbaldehyde,3-methyl-1h-pyrazole-4-carboxaldehyde,3-methylpyrazole-4-carboxaldehyde,3-methylpyrazole-4-carbaldehyde,1h-pyrazole-4-carboxaldehyde, 3-methyl,3-methyl-2h-pyrazole-4-carbaldehyde,5-methylpyrazole-4-carbaldehyde,pubchem16065,acmc-1brby,4-formyl-3-methylpyrazole PubChem CID: 2760056 IUPAC Name: 5-methyl-1H-pyrazole-4-carbaldehyde SMILES: CC1=C(C=O)C=NN1