accessibility menu, dialog, popup

UserName

CAS RN 112372-14-2

OOHON

CAS RN 112372-14-2

IUPAC Name: furo[3,2-b]pyridine-2-carboxylic acid
Synonyms: furo[3,2-b]pyridine-2-carboxylic acid
Molecular Weight (g/mol): 163.13
Molecular Formula: C8H5NO3
InChi Key: VHRCHODSRLMXFM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=NC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results