CAS RN 1123-95-1
CAS RN 1123-95-1
IUPAC Name:
6-amino-5-(hydroxymethyl)-1,2-dihydropyrimidin-2-one
Synonyms:
5-hydroxymethylcytosine
Molecular Weight (g/mol):
141.13
Molecular Formula:
C5H7N3O2
InChi Key:
RYVNIFSIEDRLSJ-UHFFFAOYSA-N
SMILES:
NC1=C(CO)C=NC(=O)N1
1
–
1
of
1
results
1
5-(Hydroxymethyl)cytosine, TRC
CAS: 1123-95-1 Molecular Formula: C5 H7 N3 O2 Molecular Weight (g/mol): 141.13 Synonym: 6-Amino-5-(hydroxymethyl)-2(1H)-pyrimidinone; IUPAC Name: 4-amino-5-(hydroxymethyl)pyrimidin-2-ol SMILES: Nc1nc(O)ncc1CO