accessibility menu, dialog, popup

UserName

CAS RN 112275-50-0

CH3H3CCH3OONHN

CAS RN 112275-50-0

IUPAC Name: tert-butyl 1,4-diazepane-1-carboxylate
Synonyms: tert-butyl 1,4-diazepane-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: WDPWEXWMQDRXAL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCCNCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)