accessibility menu, dialog, popup

UserName

H3CSSH

CAS RN 1122-97-0

IUPAC Name: 4-(methylsulfanyl)benzene-1-thiol
Synonyms: 4-(methylsulfanyl)benzenethiol
Molecular Weight (g/mol): 156.26
Molecular Formula: C7H8S2
InChi Key: KYMOWQQIZINTJZ-UHFFFAOYSA-N
SMILES: CSC1=CC=C(S)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)