accessibility menu, dialog, popup

UserName

H3CFClOH

CAS RN 112108-68-6

IUPAC Name: 1-(2-chloro-4-fluorophenyl)ethan-1-ol
Synonyms: 1-(2-chloro-4-fluorophenyl)ethanol
Molecular Weight (g/mol): 174.6
Molecular Formula: C8H8ClFO
InChi Key: UVTJHIWKGYOZFO-UHFFFAOYNA-N
SMILES: CC(O)C1=C(Cl)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results