accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 1120-28-1

IUPAC Name: methyl icosanoate
Synonyms: eicosanoic acid, methyl ester
Molecular Weight (g/mol): 326.57
Molecular Formula: C21H42O2
InChi Key: QGBRLVONZXHAKJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)