accessibility menu, dialog, popup

UserName

CAS RN 112-04-9

H3CSiClClCl

CAS RN 112-04-9

IUPAC Name: trichloro(octadecyl)silane
Synonyms: octadecyltrichlorosilane
Molecular Weight (g/mol): 387.93
Molecular Formula: C18H37Cl3Si
InChi Key: PYJJCSYBSYXGQQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)