CAS RN 111974-73-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Phenyl N-[(2-Phenylsulfanyl)phenyl]carbamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Phenyl[2-phenylthio)phenyl]carbamate, TRC
CAS: 111974-73-3 Molecular Formula: C19 H15 N O2 S Molecular Weight (g/mol): 321.39 Synonym: Carbamic acid, N-[2-(phenylthio)phenyl]-, phenyl ester,Carbamic acid, [2-(phenylthio)phenyl]-, phenyl ester (9CI),Phenyl [2-(phenylthio)phenyl]carbamate,Phenyl N-[(2-Phenylsulfanyl)phenyl]carbamate,N-[2-(Phenylthio)phenyl]carbamic Acid Phenyl Ester IUPAC Name: phenyl N-(2-phenylsulfanylphenyl)carbamate SMILES: O=C(Nc1ccccc1Sc2ccccc2)Oc3ccccc3