accessibility menu, dialog, popup

UserName

CAS RN 111860-00-5

CH3SBr

CAS RN 111860-00-5

IUPAC Name: 2-bromo-5-methyl-1-benzothiophene
Synonyms: 2-bromo-5-methyl-1-benzothiophene
Molecular Weight (g/mol): 227.12
Molecular Formula: C9H7BrS
InChi Key: HZPBPLLBZYSGBI-UHFFFAOYSA-N
SMILES: CC1=CC=C2SC(Br)=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results