accessibility menu, dialog, popup

UserName

CH3NOSOOFFF

CAS RN 111770-91-3

IUPAC Name: 6-methylpyridin-3-yl trifluoromethanesulfonate
Synonyms: 6-methylpyridin-3-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 241.18
Molecular Formula: C7H6F3NO3S
InChi Key: NSSSVJYQTZRVMH-UHFFFAOYSA-N
SMILES: CC1=NC=C(OS(=O)(=O)C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results