accessibility menu, dialog, popup

UserName

CH3SOOOHONH2

CAS RN 111769-27-8

IUPAC Name: 4-methylbenzene-1-sulfonic acid; oxolan-3-amine
Synonyms: oxolan-3-amine; para-toluene sulfonate
Molecular Weight (g/mol): 259.32
Molecular Formula: C11H17NO4S
InChi Key: BZXPLADBSZWDIH-UHFFFAOYNA-N
SMILES: NC1CCOC1.CC1=CC=C(C=C1)S(O)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results