accessibility menu, dialog, popup

UserName

H3CSNHCH3OOHO

CAS RN 1115-47-5

IUPAC Name: (2R)-2-acetamido-4-(methylsulfanyl)butanoate
Synonyms: (2R)-2-acetamido-4-(methylsulfanyl)butanoate
Molecular Weight (g/mol): 190.24
Molecular Formula: C7H12NO3S
InChi Key: XUYPXLNMDZIRQH-ZCFIWIBFSA-M
SMILES: CSCC[C@@H](NC(C)=O)C([O-])=O
Molecular Weight (g/mol) 
  • (9)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (9)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)