accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CFO

CAS RN 1112209-24-1

IUPAC Name: 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Synonyms: 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Molecular Weight (g/mol): 250.08
Molecular Formula: C13H16BFO3
InChi Key: WENPYTYJEVDZDV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(C=O)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results