accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNNH2CH3

CAS RN 1111637-91-2

IUPAC Name: 3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms: 3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol): 234.11
Molecular Formula: C12H19BN2O2
InChi Key: BWEOUBYDXFBFMA-UHFFFAOYSA-N
SMILES: CC1=C(N)N=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results