CAS RN 111-96-6
Bis(2-methoxyethyl) ether, 99%, extra pure
CAS: 111-96-6 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.18 InChI Key: SBZXBUIDTXKZTM-UHFFFAOYSA-N Synonym: diglyme,bis 2-methoxyethyl ether,2-methoxyethyl ether,1-methoxy-2-2-methoxyethoxy ethane,diethylene glycol dimethyl ether,poly-solv,di 2-methoxyethyl ether,dimethyl carbitol,2,5,8-trioxanonane PubChem CID: 8150 ChEBI: CHEBI:46784 IUPAC Name: 1-methoxy-2-(2-methoxyethoxy)ethane SMILES: COCCOCCOC
Bis(2-methoxyethyl) ether, 99+%, Extra Dry, AcroSeal™
CAS: 111-96-6 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.18 MDL Number: MFCD00008503 InChI Key: SBZXBUIDTXKZTM-UHFFFAOYSA-N Synonym: diglyme,bis 2-methoxyethyl ether,2-methoxyethyl ether,1-methoxy-2-2-methoxyethoxy ethane,diethylene glycol dimethyl ether,poly-solv,di 2-methoxyethyl ether,dimethyl carbitol,2,5,8-trioxanonane PubChem CID: 8150 ChEBI: CHEBI:46784 IUPAC Name: 1-methoxy-2-(2-methoxyethoxy)ethane SMILES: COCCOCCOC
Diethylene glycol dimethyl ether, 99%, stab. with 100ppm BHT
CAS: 111-96-6 Molecular Formula: C6H14O3 MDL Number: MFCD00008503 InChI Key: SBZXBUIDTXKZTM-UHFFFAOYSA-N Synonym: diglyme,bis 2-methoxyethyl ether,2-methoxyethyl ether,1-methoxy-2-2-methoxyethoxy ethane,diethylene glycol dimethyl ether,poly-solv,di 2-methoxyethyl ether,dimethyl carbitol,2,5,8-trioxanonane PubChem CID: 8150 ChEBI: CHEBI:46784 IUPAC Name: 1-methoxy-2-(2-methoxyethoxy)ethane SMILES: COCCOCCOC
Bis(2-methoxyethyl) ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™
CAS: 111-96-6 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.18 InChI Key: SBZXBUIDTXKZTM-UHFFFAOYSA-N Synonym: diglyme,bis 2-methoxyethyl ether,2-methoxyethyl ether,1-methoxy-2-2-methoxyethoxy ethane,diethylene glycol dimethyl ether,poly-solv,di 2-methoxyethyl ether,dimethyl carbitol,2,5,8-trioxanonane PubChem CID: 8150 ChEBI: CHEBI:46784 IUPAC Name: 1-methoxy-2-(2-methoxyethoxy)ethane SMILES: COCCOCCOC
Bis(2-methoxyethyl) ether, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
Diethylene Glycol Dimethyl Ether, 99%, Reagent Grade, Honeywell™
CAS: 111-96-6 Molecular Formula: C6H14O3 Molecular Weight (g/mol): 134.175 MDL Number: MFCD00008503 InChI Key: SBZXBUIDTXKZTM-UHFFFAOYSA-N Synonym: diglyme,bis 2-methoxyethyl ether,2-methoxyethyl ether,1-methoxy-2-2-methoxyethoxy ethane,diethylene glycol dimethyl ether,poly-solv,di 2-methoxyethyl ether,dimethyl carbitol,2,5,8-trioxanonane PubChem CID: 8150 ChEBI: CHEBI:46784 IUPAC Name: 1-methoxy-2-(2-methoxyethoxy)ethane SMILES: COCCOCCOC