accessibility menu, dialog, popup

UserName

H3COH

CAS RN 111-87-5

IUPAC Name: octan-1-ol
Synonyms: octanol caprylic alcohol (substance) lorol C8 alcohol C-8 lorol 20 fema number 2800 capryl alcohol octyl alcohol, primary 1-oktanol prim-n-octyl alcohol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: KBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILES: CCCCCCCCO
Quantity 
  • (7)
  • (1)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (6)
  • (8)
  • (3)
  • (6)
  • (6)
  • (2)
Percent Purity 
  • (7)
  • (1)
  • (3)
  • (11)
Grade 
  • (2)
  • (5)
  • (1)
  • (4)
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (4)
  • (1)
  • (2)
  • (6)
  • (6)
  • (3)
  • (2)
  • (7)
  • (1)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (6)
  • (8)
  • (3)
  • (6)
  • (6)
  • (2)
  • (7)
  • (1)
  • (3)
  • (11)
  • (2)
  • (5)
  • (1)
  • (4)
  • (2)
  • (6)
  • (19)
  • (1)