accessibility menu, dialog, popup

UserName

CAS RN 111-49-9

HN

CAS RN 111-49-9

IUPAC Name: azepane
Synonyms: hexamethyleneimine
Molecular Weight (g/mol): 99.18
Molecular Formula: C6H13N
InChi Key: ZSIQJIWKELUFRJ-UHFFFAOYSA-N
SMILES: C1CCCNCC1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)