accessibility menu, dialog, popup

UserName

HOOCH2

CAS RN 111-45-5

IUPAC Name: 2-(prop-2-en-1-yloxy)ethan-1-ol
Synonyms: peg-12 allyl ether
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: GCYHRYNSUGLLMA-UHFFFAOYSA-N
SMILES: OCCOCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results