accessibility menu, dialog, popup

UserName

H3CN

CAS RN 110-59-8

IUPAC Name: pentanenitrile
Synonyms: valeronitrile
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: RFFFKMOABOFIDF-UHFFFAOYSA-N
SMILES: CCCCC#N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)