accessibility menu, dialog, popup

UserName

CAS RN 110-21-4

IUPAC Name: (carbamoylamino)urea
Synonyms: 1,2-hydrazinedicarboxamide
Molecular Weight (g/mol): 118.1
Molecular Formula: C2H6N4O2
InChi Key: ULUZGMIUTMRARO-UHFFFAOYSA-N
SMILES: NC(=O)NNC(N)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results