accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 110-13-4

IUPAC Name: hexane-2,5-dione
Synonyms: 2,5-hexanedione
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: OJVAMHKKJGICOG-UHFFFAOYSA-N
SMILES: CC(=O)CCC(C)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)