CAS RN 109806-71-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(p-Chloro-alpha-phenylbenzyl)-1-piperazineethanol, TRC
CAS: 109806-71-5 Molecular Formula: C19 H23 Cl N2 O Molecular Weight (g/mol): 330.85 Synonym: 1-Piperazineethanol, 4-[(4-chlorophenyl)phenylmethyl]-,1-Piperazineethanol, 4-(p-chloro-α-phenylbenzyl)- (6CI),4-[(4-Chlorophenyl)phenylmethyl]-1-piperazineethanol,2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol,2-[4-[(RS)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol IUPAC Name: 2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethanol SMILES: OCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3