accessibility menu, dialog, popup

UserName

CAS RN 1093860-45-7

H3CNNNH2HClHCl

CAS RN 1093860-45-7

IUPAC Name: 1-(1-methyl-1H-indazol-3-yl)methanamine dihydrochloride
Synonyms: 1-(1-methylindazol-3-yl)methanamine dihydrochloride
Molecular Weight (g/mol): 234.12
Molecular Formula: C9H13Cl2N3
InChi Key: ZHWICCCSNYFLJY-UHFFFAOYSA-N
SMILES: Cl.Cl.CN1N=C(CN)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results