accessibility menu, dialog, popup

UserName

CAS RN 1092461-35-2

IUPAC Name: 2-[3-bromo-5-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [3-bromo-5-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 299.04
Molecular Formula: C9H6BrF3O3
InChi Key: PEBPBUMXKAQCGP-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(OC(F)(F)F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results