accessibility menu, dialog, popup

UserName

CAS RN 1092461-15-8

ClOOFFF

CAS RN 1092461-15-8

IUPAC Name: 5-chloro-2-(trifluoromethoxy)benzaldehyde
Synonyms: 5-chloro-2-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: VJRCCSALVWMZAN-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(C=O)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results