accessibility menu, dialog, popup

UserName

CAS RN 109-97-7

IUPAC Name: 1H-pyrrole
Synonyms: pyrrole divinyleneimine divinylenimine parzate
Molecular Weight (g/mol): 67.09
Molecular Formula: C4H5N
InChi Key: KAESVJOAVNADME-UHFFFAOYSA-N
SMILES: N1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (7)
  • (4)