accessibility menu, dialog, popup

UserName

CH3OHO

CAS RN 109-86-4

IUPAC Name: 2-methoxyethan-1-ol
Synonyms: 2-methoxyethanol MECS polyoxyethylene (40) monomethyl ether caswell no. 551 polyethylene glycol 300 methyl ether calcium salt methyl cellosolve EGME methylcelosolv methoxypolyethylene glycol polyethylene glycol 2000 monomethyl ether
Molecular Weight (g/mol): 76.1
Molecular Formula: C3H8O2
InChi Key: XNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILES: COCCO
Molecular Weight (g/mol) 
  • (18)
  • (25)
Quantity 
  • (13)
  • (1)
  • (2)
  • (9)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
Boiling Point 
  • (9)
  • (10)
  • (13)
  • (2)
Grade 
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
Physical Form 
  • (26)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (11)
  • (10)
  • (3)
  • (2)
  • (18)
  • (25)
  • (13)
  • (1)
  • (2)
  • (9)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
  • (9)
  • (10)
  • (13)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (26)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)