accessibility menu, dialog, popup

UserName

H3COClO

CAS RN 109-61-5

IUPAC Name: propyl carbonochloridate
Synonyms: carbonochloridic acid, propyl ester
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: QQKDTTWZXHEGAQ-UHFFFAOYSA-N
SMILES: CCCOC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results