accessibility menu, dialog, popup

UserName

CAS RN 109-16-0

H3CCH2OOOOOOCH3CH2

CAS RN 109-16-0

IUPAC Name: 2-(2-{2-[(2-methylprop-2-enoyl)oxy]ethoxy}ethoxy)ethyl 2-methylprop-2-enoate
Synonyms: triethyleneglycol dimethacrylate triethylene glycol dimethacrylate
Molecular Weight (g/mol): 286.32
Molecular Formula: C14H22O6
InChi Key: HWSSEYVMGDIFMH-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCOCCOCCOC(=O)C(C)=C
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results