accessibility menu, dialog, popup

UserName

CAS RN 108-48-5

IUPAC Name: 2,6-dimethylpyridine
Synonyms: lutidine 2,6-dimethypyridine lut α,α'-lutidine hsdb 79
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: OISVCGZHLKNMSJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (7)
Boiling Point 
  • (4)
  • (3)
  • (1)
Percent Purity 
  • (4)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (7)
  • (4)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)