accessibility menu, dialog, popup

UserName

CAS RN 108-13-4

IUPAC Name: propanediamide
Synonyms: propanediamide
Molecular Weight (g/mol): 102.09
Molecular Formula: C3H6N2O2
InChi Key: WRIRWRKPLXCTFD-UHFFFAOYSA-N
SMILES: NC(=O)CC(N)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results