CAS RN 107880-74-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Famotidine Acid, TRC
CAS: 107880-74-0 Molecular Formula: C8 H12 N4 O2 S2 Molecular Weight (g/mol): 260.34 Synonym: Propanoic acid, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-;Famotidine Acid Impurity,3-[[[2-[(Diaminomethylene)-amino]thiazol-4-yl]methyl]sulfanyl]-propanoic Acid,Famotidine Imp. F (EP) IUPAC Name: 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanoic acid SMILES: NC(=Nc1nc(CSCCC(=O)O)cs1)N
2
3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.