CAS RN 107754-14-3
CAS RN 107754-14-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 4-[(5-Amino-1-methylindol-3-yl)methyl]-3-methoxybenzoate, TRC
CAS: 107754-14-3 Molecular Formula: C19 H20 N2 O3 Molecular Weight (g/mol): 324.37 Synonym: 4-[(5-Amino-1-methyl-1H-indol-3-yl)methyl]-3-methoxy-benzoic Acid Methyl Ester IUPAC Name: methyl 4-[(5-amino-1-methyl-indol-3-yl)methyl]-3-methoxy-benzoate SMILES: COC(=O)c1ccc(Cc2cn(C)c3ccc(N)cc23)c(OC)c1