accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNSOH

CAS RN 1073371-73-9

IUPAC Name: [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl](1,3-thiazol-2-yl)methanol
Synonyms: [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl](1,3-thiazol-2-yl)methanol
Molecular Weight (g/mol): 317.21
Molecular Formula: C16H20BNO3S
InChi Key: WLBDZEXNSNOBAQ-UHFFFAOYNA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(O)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results