accessibility menu, dialog, popup

UserName

CH3CH3I

CAS RN 107-08-4

IUPAC Name: 2-iodopropane
Synonyms: 2-iodopropane
Molecular Weight (g/mol): 169.99
Molecular Formula: C3H7I
InChi Key: FMKOJHQHASLBPH-UHFFFAOYSA-N
SMILES: CC(C)I
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)