CAS RN 1068-42-4
IUPAC Name:
N''-aminoguanidine; sulfuric acid
Synonyms:
amino-guanidine; sulfuric acid
Molecular Weight (g/mol):
172.16
Molecular Formula:
CH8N4O4S
InChi Key:
XLONNWGCEFSFTN-UHFFFAOYSA-N
SMILES:
NN=C(N)N.OS(O)(=O)=O
1
–
1
of
1
results
1
Alfa Aesar™ Aminoguanidine sulfate, 98%
CAS: 1068-42-4 Molecular Formula: CH8N4O4S Molecular Weight (g/mol): 172.16 MDL Number: MFCD00151369 InChI Key: XLONNWGCEFSFTN-UHFFFAOYSA-N Synonym: aminoguanidine sulfate,guanylhydrazinesulfate,aminoguanidine sulphate,carbazamidine sulfate,2-aminoguanidine; sulfuric acid,amino-guanidine; sulfuric acid,aminoguanidinium sulfate,1-aminoguanidinium sulfate,ch6n4.h2so4,spectrum1505162 PubChem CID: 114832 IUPAC Name: 2-aminoguanidine;sulfuric acid SMILES: NN=C(N)N.OS(O)(=O)=O