accessibility menu, dialog, popup

UserName

NaCHC

CAS RN 1066-26-8

IUPAC Name: sodium acetylene
Synonyms: monosodium acetylide
Molecular Weight (g/mol): 48.02
Molecular Formula: C2HNa
InChi Key: SFDZETWZUCDYMD-UHFFFAOYSA-N
SMILES: [Na+].C#[C-]
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)