CAS RN 106040-48-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tribenuron, TRC
CAS: 106040-48-6 Molecular Formula: C14H15N5O6S Molecular Weight (g/mol): 381.36 Synonym: 2-[[[[(4-Methoxy-6-methyl-1,3,5-triazin-2-yl)methylamino]carbonyl]amino]sulfonyl]-benzoic Acid; IUPAC Name: 2-(N-((4-methoxy-6-methyl-1,3,5-triazin-2-yl)(methyl)carbamoyl)sulfamoyl)benzoic acid SMILES: O=C(O)C1=CC=CC=C1S(=O)(NC(N(C2=NC(OC)=NC(C)=N2)C)=O)=O