accessibility menu, dialog, popup

UserName

H3COCH3

CAS RN 106-68-3

IUPAC Name: octan-3-one
Synonyms: 3-octanone
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: RHLVCLIPMVJYKS-UHFFFAOYSA-N
SMILES: CCCCCC(=O)CC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results