CAS RN 105956-99-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Clinafloxacin Hydrochloride, TRC
CAS: 105956-99-8 Molecular Formula: C17 H17 Cl F N3 O3 . Cl H Molecular Weight (g/mol): 402.25 Synonym: Clinafloxacin Hydrochloride,3-Quinolinecarboxylic acid, 7-(3-amino-1-pyrrolidinyl)-8-chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-, monohydrochloride (9CI),AM 1091,CI 960,Clinafloxacin hydrochloride,KR 11095,3-Quinolinecarboxylic acid, 7-(3-amino-1-pyrrolidinyl)-8-chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-, hydrochloride (1:1) IUPAC Name: 7-(3-aminopyrrolidin-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxoquinoline-3-carboxylic acid;hydrochloride SMILES: Cl.NC1CCN(C1)c2c(F)cc3C(=O)C(=CN(C4CC4)c3c2Cl)C(=O)O