accessibility menu, dialog, popup

Test

CAS RN 10593-29-0

CAS RN 10593-29-0

IUPAC Name: sodium [(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanide
Synonyms: sodium [(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanide
Molecular Weight (g/mol): 218.2
Molecular Formula: C6H11NaO5S
InChi Key: VKPBZIVFRYLHPT-WNFIKIDCSA-M
SMILES: [Na+].OC[C@H]1O[C@@H]([S-])[C@H](O)[C@@H](O)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results