accessibility menu, dialog, popup

UserName

ONH2HCl

CAS RN 10578-75-3

IUPAC Name: 2-(benzyloxy)ethan-1-amine hydrochloride
Synonyms: 2-(benzyloxy)ethanamine hydrochloride
Molecular Weight (g/mol): 187.67
Molecular Formula: C9H14ClNO
InChi Key: AFQMWBUNDONXED-UHFFFAOYSA-N
SMILES: Cl.NCCOCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results