accessibility menu, dialog, popup

UserName

CAS RN 105219-44-1

H3COFOHOF

CAS RN 105219-44-1

IUPAC Name: 3-(3,5-difluoro-4-methoxyphenyl)propanoic acid
Synonyms: 3-(3,5-difluoro-4-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 216.18
Molecular Formula: C10H10F2O3
InChi Key: WOUCCZYVNXELFX-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(CCC(O)=O)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results